ENA321764660 Enamine

1-{3-methyl-[1,2,4]triazolo[4,3-b]pyridazin-6-yl}piperazine dihydrochloride

  • MDL number MFCD18839022
  • Cas Number : 1306605-44-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-77538

Safety Information

msds https://www.enaminestore.com/catalog/EN300-77538
Availability details for
Quantity
  • Availability
  • Add to Cart