ENA321764674 Enamine

4-(propan-2-yl)azepane

  • MDL number MFCD18839038
  • Cas Number : 1306606-61-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-77619

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-77619
Availability details for
Quantity
  • Availability
  • Add to Cart