ENA321764680 Enamine

tert-butyl N-{2-[(hydrazinecarbonyl)amino]ethyl}carbamate

  • MDL number MFCD18917281
  • Cas Number : 1311314-62-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-77654

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-77654
Availability details for
Quantity
  • Availability
  • Add to Cart