ENA321764683 Enamine

3-[4-(trifluoromethyl)-1H-pyrazol-1-yl]aniline

  • MDL number MFCD18839046
  • Cas Number : 1306605-68-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-77665

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-77665
Availability details for
Quantity
  • Availability
  • Add to Cart