ENA321764685 Enamine

N-methyl-1H-imidazole-4-sulfonamide

  • MDL number MFCD18839048
  • Cas Number : 58768-15-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 157-158
Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-77672

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-77672
Availability details for
Quantity
  • Availability
  • Add to Cart