ENA321764686 Enamine

N,N-dimethyl-1H-imidazole-4-sulfonamide

  • MDL number MFCD18839049
  • Cas Number : 58768-16-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-77673

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-77673
Availability details for
Quantity
  • Availability
  • Add to Cart