ENA321764707 Enamine

4-(2-amino-1,3-thiazol-4-yl)-5-methylfuran-2-carboxamide

  • MDL number MFCD18839071
  • Cas Number : 1306605-46-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-77715

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-77715
Availability details for
Quantity
  • Availability
  • Add to Cart