ENA321764710 Enamine

ethyl 5-bromo-2H-indazole-3-carboxylate

  • MDL number MFCD18839073
  • Cas Number : 1081-04-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-77718

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-77718
Availability details for
Quantity
  • Availability
  • Add to Cart