ENA321764712 Enamine

pent-3-en-1-amine hydrochloride

  • Cas Number : 1306615-57-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-77728

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-77728
Availability details for
Quantity
  • Availability
  • Add to Cart