ENA321764713 Enamine

tert-butyl N-[(N'-hydroxycarbamimidoyl)(2-methoxyphenyl)methyl]carbamate

  • Cas Number : 1306615-56-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-77731

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-77731
Availability details for
Quantity
  • Availability
  • Add to Cart