ENA321764714 Enamine

2-[3-(3-chlorophenyl)-1,2,4-oxadiazol-5-yl]ethan-1-amine hydrochloride

  • MDL number MFCD18839077
  • Cas Number : 1306605-82-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-77732

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-77732
Availability details for
Quantity
  • Availability
  • Add to Cart