ENA321764718 Enamine

2-[(4-methylphenyl)methyl]pyrrolidine

  • MDL number MFCD02663496
  • Cas Number : 383127-62-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-76684

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-76684
Availability details for
Quantity
  • Availability
  • Add to Cart