ENA321764719 Enamine

2-(2-iodophenyl)pyrrolidine

  • MDL number MFCD02663434
  • Cas Number : 383127-23-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-76685

Safety Information

msds https://www.enaminestore.com/catalog/EN300-76685
Availability details for
Quantity
  • Availability
  • Add to Cart