ENA321764721 Enamine

(2S)-N-(6-methylpyridin-2-yl)pyrrolidine-2-carboxamide

  • MDL number MFCD09723799
  • Cas Number : 478912-51-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 62...64
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-76131

Safety Information

msds https://www.enaminestore.com/catalog/EN300-76131
Availability details for
Quantity
  • Availability
  • Add to Cart