ENA321764730 Enamine

3-fluoro-4-{[5-(trifluoromethyl)pyridin-2-yl]oxy}aniline

  • MDL number MFCD12450702
  • Cas Number : 1094937-85-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-76701

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-76701
Availability details for
Quantity
  • Availability
  • Add to Cart