ENA321764735 Enamine

1-[(pyridin-3-yl)methyl]piperidin-4-amine trihydrochloride

  • MDL number MFCD08689422
  • Cas Number : 1185293-43-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 3HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-76706

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-76706
Availability details for
Quantity
  • Availability
  • Add to Cart