ENA321764739 Enamine

pyrrolidin-2-ylmethanesulfonamide

  • MDL number MFCD14688721
  • Cas Number : 1247347-73-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 72...74
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-75954

Safety Information

signalword Warning
msds https://www.enaminestore.com/catalog/EN300-75954
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart