ENA321764741 Enamine

3-(pyrrolidine-1-sulfonyl)benzene-1-sulfonyl chloride

  • MDL number MFCD08444952
  • Cas Number : 920449-65-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-75956

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-75956
Availability details for
Quantity
  • Availability
  • Add to Cart