ENA321764748 Enamine

2-(4-cyclohexanecarbonylpiperazin-1-yl)ethan-1-amine

  • MDL number MFCD10015952
  • Cas Number : 1018552-50-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-76007

Safety Information

msds https://www.enaminestore.com/catalog/EN300-76007
Availability details for
Quantity
  • Availability
  • Add to Cart