ENA321764749 Enamine

2-(2-chloro-4-fluorophenyl)-2-methylpropan-1-amine

  • MDL number MFCD17282027
  • Cas Number : 1004285-25-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-76008

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-76008
Availability details for
Quantity
  • Availability
  • Add to Cart