ENA321764751 Enamine

1-(6-ethylpyrimidin-4-yl)piperidin-3-amine

  • MDL number MFCD14617978
  • Cas Number : 1251240-01-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-76156

Safety Information

msds https://www.enaminestore.com/catalog/EN300-76156
Availability details for
Quantity
  • Availability
  • Add to Cart