ENA321764754 Enamine

1-[(2-chloropyridin-3-yl)sulfonyl]-4-methylpiperazine

  • MDL number MFCD07367060
  • Cas Number : 60597-73-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-76162

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-76162
Availability details for
Quantity
  • Availability
  • Add to Cart