ENA321764758 Enamine

N'-hydroxy-2-(1H-pyrazol-1-yl)ethanimidamide

  • Cas Number : 1158119-05-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-76745

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-76745
Availability details for
Quantity
  • Availability
  • Add to Cart