ENA321764763 Enamine

2-[(dimethylamino)methyl]-4-methylaniline

  • MDL number MFCD11156628
  • Cas Number : 1021144-66-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-76751

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-76751
Availability details for
Quantity
  • Availability
  • Add to Cart