ENA321764767 Enamine

1-(1-bromoethyl)-3,5-bis(trifluoromethyl)benzene

  • MDL number MFCD18762083
  • Cas Number : 160376-84-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. -20
Shipping Temperature Ice Pack
Product Link https://www.enaminestore.com/catalog/EN300-76755

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-76755
Availability details for
Quantity
  • Availability
  • Add to Cart