ENA321764778 Enamine

2,3-dimethylbutanoic acid

  • MDL number MFCD00045888
  • Cas Number : 14287-61-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-76184

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-76184
Availability details for
Quantity
  • Availability
  • Add to Cart