ENA321764782 Enamine

5-(chloromethyl)-3-phenyl-1,3-oxazolidin-2-one

  • MDL number MFCD01024968
  • Cas Number : 711-85-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 103...105
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-76024

Safety Information

signalword Danger
msds https://www.enaminestore.com/catalog/EN300-76024
pictograms GHS05,GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart