ENA321764783 Enamine

N-methylpiperidine-3-sulfonamide

  • MDL number MFCD09951058
  • Cas Number : 1017026-28-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-76772

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-76772
Availability details for
Quantity
  • Availability
  • Add to Cart