ENA321764784 Enamine

2-(cyclopentyloxy)butanoic acid

  • MDL number MFCD12067173
  • Cas Number : 1157982-27-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-76025

Safety Information

msds https://www.enaminestore.com/catalog/EN300-76025
Availability details for
Quantity
  • Availability
  • Add to Cart