ENA321764785 Enamine

4-(chloromethyl)-2-methylquinoline

  • MDL number MFCD06797694
  • Cas Number : 288399-19-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-76774

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-76774
Availability details for
Quantity
  • Availability
  • Add to Cart