ENA321764788 Enamine

benzyl N-(1-phenylbut-3-en-1-yl)carbamate

  • MDL number MFCD00973293
  • Cas Number : 138723-71-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-76777

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-76777
Availability details for
Quantity
  • Availability
  • Add to Cart