ENA321764790 Enamine

2-[butyl(methyl)amino]-5-(trifluoromethyl)benzoic acid

  • MDL number MFCD17114491
  • Cas Number : 1306606-00-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-76779

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-76779
Availability details for
Quantity
  • Availability
  • Add to Cart