ENA321764791 Enamine

1-(aminomethyl)-N,N-dimethylcyclopentane-1-carboxamide

  • MDL number MFCD16118419
  • Cas Number : 1251394-34-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-76780

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-76780
Availability details for
Quantity
  • Availability
  • Add to Cart