ENA321764801 Enamine

1-(2-bromo-5-fluorophenyl)propan-2-one

  • MDL number MFCD06797890
  • Cas Number : 1250521-52-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-76796

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-76796
Availability details for
Quantity
  • Availability
  • Add to Cart