ENA321764803 Enamine

2-[(2-fluorophenyl)sulfanyl]ethan-1-amine

  • MDL number MFCD09936291
  • Cas Number : 1016891-91-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-76798

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-76798
Availability details for
Quantity
  • Availability
  • Add to Cart