ENA321764807 Enamine

methyl 3-(4-hydroxypiperidin-1-yl)propanoate

  • MDL number MFCD11174791
  • Cas Number : 147023-16-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-76029

Safety Information

signalword Danger
msds https://www.enaminestore.com/catalog/EN300-76029
pictograms GHS05,GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart