ENA321764808 Enamine

2-cyclopentyl-2-hydroxyacetic acid

  • MDL number MFCD16669022
  • Cas Number : 6053-71-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-76030

Safety Information

msds https://www.enaminestore.com/catalog/EN300-76030
Availability details for
Quantity
  • Availability
  • Add to Cart