ENA321764809 Enamine

[5-bromo-2-(morpholin-4-yl)pyridin-3-yl]methanol

  • MDL number MFCD16159504
  • Cas Number : 1247213-10-0
  • Molecular Weight : 273.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-75949

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-75949
Availability details for
Quantity
  • Availability
  • Add to Cart