ENA321764810 Enamine

1-amino-2-cyclopentylpropan-2-ol

  • MDL number MFCD16830668
  • Cas Number : 1249360-64-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-76216

Safety Information

msds https://www.enaminestore.com/catalog/EN300-76216
Availability details for
Quantity
  • Availability
  • Add to Cart