ENA321764813 Enamine

2-(phenylformamido)-3-(pyridin-3-yl)prop-2-enoic acid

  • Cas Number : 33560-99-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-76034

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-76034
Availability details for
Quantity
  • Availability
  • Add to Cart