ENA321764816 Enamine

1-(pyridine-3-carbonyl)-2,3-dihydro-1H-indol-5-amine

  • MDL number MFCD09261832
  • Cas Number : 927996-80-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-76816

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-76816
Availability details for
Quantity
  • Availability
  • Add to Cart