ENA321764821 Enamine

4-(5-amino-1H-pyrazol-1-yl)benzonitrile

  • MDL number MFCD11641451
  • Cas Number : 1152964-72-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-76037

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-76037
Availability details for
Quantity
  • Availability
  • Add to Cart