ENA321764835 Enamine

(3S)-3-amino-3-(5-methylfuran-2-yl)propanoic acid

  • MDL number MFCD12797111
  • Cas Number : 1269994-47-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-76269

Safety Information

msds https://www.enaminestore.com/catalog/EN300-76269
Availability details for
Quantity
  • Availability
  • Add to Cart