ENA321764838 Enamine

4-(cyclopropylmethoxy)aniline hydrochloride

  • MDL number MFCD15200817
  • Cas Number : 1158573-73-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-76276

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-76276
Availability details for
Quantity
  • Availability
  • Add to Cart