ENA321764840 Enamine

ethyl 2-(2-aminophenyl)acetate hydrochloride

  • MDL number MFCD06659777
  • Cas Number : 61-88-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 117...119
Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-76861

Safety Information

msds https://www.enaminestore.com/catalog/EN300-76861
Availability details for
Quantity
  • Availability
  • Add to Cart