ENA321764843 Enamine

[5-bromo-2-(3-ethylpiperidin-1-yl)pyridin-3-yl]methanol

  • MDL number MFCD15503844
  • Cas Number : 1306603-66-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-76864

Safety Information

msds https://www.enaminestore.com/catalog/EN300-76864
Availability details for
Quantity
  • Availability
  • Add to Cart