ENA321764845 Enamine

3-(2-aminobutyl)-5,5-dimethylimidazolidine-2,4-dione

  • MDL number MFCD18838902
  • Cas Number : 1266972-99-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-76866

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-76866
Availability details for
Quantity
  • Availability
  • Add to Cart