ENA321764861 Enamine

4-(aminomethyl)-N,N-dimethyloxan-4-amine

  • MDL number MFCD09065027
  • Cas Number : 176445-80-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-76905

Safety Information

signalword Danger
msds https://www.enaminestore.com/catalog/EN300-76905
pictograms GHS05,GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart