ENA321764863 Enamine

1-phenyl-1H-imidazole-5-carboxamide

  • MDL number MFCD11643420
  • Cas Number : 1098355-91-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-76282

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-76282
Availability details for
Quantity
  • Availability
  • Add to Cart