ENA321764865 Enamine

2,2,2-trifluoro-1-(1-methyl-1H-pyrazol-4-yl)ethan-1-ol

  • MDL number MFCD12787666
  • Cas Number : 1226507-40-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-76283

Safety Information

msds https://www.enaminestore.com/catalog/EN300-76283
Availability details for
Quantity
  • Availability
  • Add to Cart